missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calycosin 7-O-Beta-D-glucoside, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Calycosin 7-Glucoside |
| CAS | 20633-67-4 |
| Molecular Formula | C22 H22 O10 |
| Quantity | 2.5 mg |
| Synonym | Calycosin 7-O-beta-D-Glucopyranoside |
| InChI Formula | InChI=1S/C22H22O10/c1-29-15-5-2-10(6-14(15)24)13-9-30-16-7-11(3-4-12(16)18(13)25)31-22-21(28)20(27)19(26)17(8-23)32-22/h2-7,9,17,19-24,26-28H,8H2,1H3/t17-,19-,20+,21-,22-/m1/s1 |
| SMILES | COc1ccc(cc1O)C2=COc3cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)ccc3C2=O |
| IUPAC Name | 3-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one |
| Molecular Weight (g/mol) | 446.4 |
| Formula Weight | 446.1213 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction