missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase I Antibody (S06-7C4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19540-100ul
This item is not returnable.
View return policy
Description
CaM Kinase I Monoclonal antibody specifically detects CaM Kinase I in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CaM Kinase I | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-7C4 | |
| Unconjugated | |
| calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
| A synthetic peptide of human CaM Kinase I (Uniprot # Q14012) | |
| 100 μg | |
| Neuroscience, Protein Kinase, Sensory Systems, Vision | |
| 8536 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction