missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase II gamma Antibody (8G10C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 196.00 - € 552.00
Specifications
| Antigen | CaM Kinase II gamma |
|---|---|
| Clone | 8G10C1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698595
|
Novus Biologicals
NBP2-37262-0.025ml |
0.025 mL |
€ 196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131079
|
Novus Biologicals
NBP2-37262 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaM Kinase II gamma Monoclonal specifically detects CaM Kinase II gamma in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| CaM Kinase II gamma | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678 | |
| CAMK2G | |
| IgG1 | |
| Affinity Purified |
| 8G10C1 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 818 | |
| Purified recombinant fragment of human CAMK2G (AA: 322-481) expressed in E. Coli. | |
| Primary | |
| 62.6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title