Learn More
Carl Zeiss™ Camera Adapters
Product Code. 11895440
Change view
Click to view available options
Description:
60N-C 0.75 in.
60N-C 1.0 in.
P95-C 0.75 in.
Magnification Power:
0.5x
0.63x
0.65x
1.0X
Unit Size:
1 unit
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Magnification Power unitSize
11895440 60N-C 0.75 in. 0.5x Each
10337143 P95-C 0.75 in. 0.65x Each
10875741 60N-C 0.75 in. 0.63x Each
15150705 60N-C 1.0 in. 1.0X 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11895440 Supplier Carl Zeiss™ Supplier No. 4261120000000
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
C-mount camera adapter for Primovert
  • Requires Primovert
  • With a phototube C mount camera adapter
  • For Primovert
Specifications
Type Camera Adapter
Description 60N-C 0.75 in.
Magnification Power 0.5x
Quantity 1 Ea.
Size 0.75 in.