missing translation for 'onlineSavingsMsg'
Learn More

Camfetamine Hydrochloride(N-Methyl-3-phenylnorbornan-2-amine Hydrochloride), Mikromol™

Product Code. 30052299
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052299 25 mg 25mg
18051500 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052299 Supplier Mikromol Supplier No. MM0995.010025
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Camfetamine Hydrochloride(N-Methyl-3-phenylnorbornan-2-amine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Camfetamine Hydrochloride (N-Methyl-3-phenylnorbornan-2-amine Hydrochloride)
CAS 39628-11-0
Analyte or Component Names Camfetamine Hydrochloride (N-Methyl-3-phenylnorbornan-2-amine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C14 H19 N . Cl H
Certifications/Compliance ISO 9001
Synonym Camfetamine Hydrochloride, N-Methyl-3-phenylnorbornan-2-amine Hydrochloride
InChI Formula InChI=1S/C14H19N.ClH/c1-15-14-12-8-7-11(9-12)13(14)10-5-3-2-4-6-10;/h2-6,11-15H,7-9H2,1H3;1H
SMILES Cl.CNC1C2CCC(C2)C1c3ccccc3
IUPAC Name N-methyl-2-phenylbicyclo[2.2.1]heptan-3-amine;hydrochloride
Molecular Weight (g/mol) 237.77
Quantity 25 mg
Formula Weight 237.1284 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.