Learn More
Camicinal, MedChemExpress
Product Code. 16477007
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477007
Brand: Medchem Express HY10922
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Camicinal (GSK962040) is a small molecule, selective motilin receptor agonist with pEC50 of 7.9.
Camicinal (GSK962040) is a small molecule, selective motilin receptor agonist with pEC50 of 7.9.
Specifications
Chemical Name or Material Camicinal
CAS 923565-21-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H33FN4O
Quantity 5 mg
Synonym GSK962040
Solubility Information DMSO : 100 mg/mL (235.54 mM; Need ultrasonic)
SMILES O=C(N1CCC(NC2=CC=CC(F)=C2)CC1)CC3=CC=C(CN4C[C@H](C)NCC4)C=C3
Molecular Weight (g/mol) 424.55
Formula Weight 424.55
Percent Purity 95.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less
Research purposes only