Learn More
CAMK2B Monoclonal Antibody (OTI4B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808428S
Encompass_Preferred
 View more versions of this product
Product Code. 17579673
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
CAMK2B Monoclonal antibody specifically detects CAMK2B in Human samples. It is validated for Western Blot
Specifications
Specifications
CAMK2B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P41279
Mouse
Affinity chromatography
RUO
1326
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI4B12
Unconjugated
MAP3K8
Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; caM kinase II subunit beta; CAM2; CaMK II; CAMK2; CAMK2B; Camk2d; CAMKB; caMK-II subunit beta; caM-kinase II beta chain; Ck2b; hypothetical protein LOC525416; MGC29528; proline rich calmodulin-dependent protein kinase; β CaMKII; βCaMKII
Human recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8 produced in E.coli.
30 μL