missing translation for 'onlineSavingsMsg'
Learn More

CAMK2B Monoclonal Antibody (OTI8D9), TrueMAB™, OriGene

Product Code. 17569503
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17569503 30 μL 30µL
17549503 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17569503 Supplier Origene Technologies Supplier No. TA808345S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CAMK2B Monoclonal antibody specifically detects CAMK2B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CAMK2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LOC100287896
Gene Alias Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; caM kinase II subunit beta; CAM2; CaMK II; CAMK2; CAMK2B; Camk2d; CAMKB; caMK-II subunit beta; caM-kinase II beta chain; Ck2b; hypothetical protein LOC525416; MGC29528; proline rich calmodulin-dependent protein kinase; β CaMKII; βCaMKII
Host Species Mouse
Immunogen Full length human recombinant protein of human MGC12965 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100287896
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.