Learn More
CAMK2G Mouse, anti-Human, Clone: 1C12H10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68519-1-IG-20UL
 View more versions of this product
Product Code. 18084265
  • € 145.00 / 20µL
Description
The CAMK2G antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAMK2G. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The CAMK2G antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CAMK2G
1C12H10
CaM kinase II subunit gamma, CAMK, CAMK II, CaMK II subunit gamma, CAMK2G, CAMKG, FLJ16043
20 μL
818
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
150 kDa