Learn More
CAMK4 Mouse, anti-Human, Clone: 1G7A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68452-1-IG-150UL
 View more versions of this product
Product Code. 18047305
  • € 410.00 / 150µL
Description
The CAMK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAMK4. This antibody recognizes human, mouse, rat antigen. The CAMK4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CAMK4
1G7A7
CaM kinase GR, CaMK GR, CaMK IV, CAMK4
150 μL
814
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
150 kDa