missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKI gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38420-20ul
This item is not returnable.
View return policy
Description
CaMKI gamma Polyclonal antibody specifically detects CaMKI gamma in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CaMKI gamma | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| calcium/calmodulin-dependent protein kinase IG, calcium/calmodulin-dependent protein kinase type 1G, CaM kinase I gamma, CaM kinase IG, CaMKI gamma, CaM-KI gamma, caMKIG, CaMK-like CREB kinase III, CLICKIII, dJ272L16.1, EC 2.7.11, EC 2.7.11.17, VWS1CLICK III | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 217-476 of human CaMKI gamma (NP_065172.1).,, Sequence:, PPFYEETESKLFEKIKEGYYEFESPFWDDISESAKDFICHLLEKDPNERYTCEKALSHPWIDGNTALHRDIYPSVSLQIQKNFAKSKWRQAFNAAAVVHHMRKLHMNLHSPGVRPEVENRPPETQASETSRPSSPEITITEAPVLDHSVALPALTQLPCQHGRRPTAPGGRSLNCLVNGSLHISSSLVPMHQGSLAAGPCGCCSSCLNIGSKGKSSYCSEPTLLKKANKKQNFKSEVMVPVKASGSSHCRAGQTGVCLIM | |
| 20 μL | |
| Protein Kinase | |
| 57172 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction