missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (23GB1290), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

€ 186.00 - € 446.00

Specifications

Antigen CaMKI
Clone 23GB1290
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088821
GREENER_CHOICE
View Documents
Invitrogen™
MA553152
100 μL
€ 446.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088765
View Documents
Invitrogen™
MA553153
20 μL
€ 186.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaMKI Recombinant Monoclonal antibody specifically detects CaMKI in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

CaMKI
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q14012, Q63450, Q91YS8
171503, 52163, 8536
IgG
Affinity chromatography
Antibody
23GB1290
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19
A synthesized peptide derived from human CaMKI.
Primary
-20°C
CAMK1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.