missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (4P4K9)
Shop All Thermo Scientific Products

Description
Positive Samples: HepG2, THP-1, Mouse lung, Mouse liver, Mouse brain, Rat lung, Rat liver, Rat brain Immunogen sequence: MLGAVEGPRW KQAEDIRDIY DFRDVLGTGA FSEVILAEDK RTQKLVAIKC IAKEALEGKE GSMENEIAVL HKIKHPNIVA LDDIYESGGH LYLIMQLVSG.
CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Specifications
Specifications
| Antigen | CaMKI |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4P4K9 |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAMK1 |
| Gene Accession No. | Q14012, Q63450, Q91YS8 |
| Gene Alias | AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?