Learn More
Invitrogen™ CaMKII alpha/beta Monoclonal Antibody (22B1), PerCP
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545148
Description
1 μg/mL was sufficient for detection of 0.2 μg CamKII by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary. Detects phosphorylated CaMKII from rat tissues. This antibody is specific for alpha and beta subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in beta ).
Ca2+/Calmodulin-Dependent Protein Kinase II (CaM Kinase II) is a multi-functional calcium and calmodulin-dependent protein kinase that mediates cellular response to a wide variety of intercellular signals (Kennedy, 1998; Schulman and Hanson, 1993). CaM Kinase II has been shown to regulate diverse cellular functions including synaptic plasticity, neurotransmitter synthesis and release, gene expression, ion channel function, carbohydrate metabolism, cytoskeletal function, and Ca2+-homeostasis (Gleason et al., 2003; Soderling, 2000; Hudmon and Schulman, 2002). Phosphorylation of Thr286 on the kinase produces an autonomously active form of CaM Kinase II (Meng et al., 2003; Picciotto et al., 1993). Autophosphorylation of Thr305 inhibits the activity CaM Kinase II. Phosphorylation at this site appears to reduce the association of CaM Kinase II with the PSD and reduces LTP and learning (Elgersma et al., 2002).
Specifications
| CaMKII alpha/beta | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.2 | |
| P08413, P11275, P11798, P28652, Q13554, Q9UQM7 | |
| Camk2a, Camk2b | |
| Synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 22B1 | |
| PerCP | |
| Camk2b | |
| alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; caM kinase II subunit alpha; CaM kinase II subunit beta; CaMK II; Camk2a; Camk2b; Camk2d; CaMKII; caMK-II subunit alpha; caMK-II subunit beta; caM-kinase II alpha chain; caM-kinase II beta chain; Ck2b; mKIAA0968; PK2CDD; PKCCD; R74975 | |
| Mouse | |
| Protein G | |
| RUO | |
| 12322, 12323, 24245, 25400, 815, 816 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.