missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKII beta/gamma/delta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539732
This item is not returnable.
View return policy
Description
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be coexpressed in the same tissue type. CaMKIV is stimulated by Ca2+ and CaM but also requires phosphorylation by a CaMK for full activation.
Specifications
| CaMKII beta/gamma/delta | |
| Polyclonal | |
| Unconjugated | |
| CAMK2G | |
| Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108058, 12323, 12325, 171140, 24245, 24246, 816, 817, 818 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13554, Q13555, Q13557, P28652, Q6PHZ2, Q923T9, P08413, P11730, P15791 | |
| CAMK2B, CAMK2D, CAMK2G | |
| A synthetic non-phosphopeptide derived from human CaMK2-beta/gamma/sigma around the phosphorylation site of Thr287 (Q-E-TP-V-E) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction