missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKII beta/gamma/delta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536562
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 50-80 kDa. Purity is >95% by SDS-PAGE.
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be coexpressed in the same tissue type. CaMKIV is stimulated by Ca2+ and CaM but also requires phosphorylation by a CaMK for full activation.
Specifications
| CaMKII beta/gamma/delta | |
| Polyclonal | |
| Unconjugated | |
| Camk2b | |
| [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit delta; calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit beta; CaM kinase II subunit delta; caM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK1; CAMK2; Camk2b; Camk2d; CAMK2G; CAMKB; CAMKD; CAMKG; Camki; CAMK-II; CaMK-II delta subunit; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; caM-kinase II beta chain; caM-kinase II delta chain; caM-kinase II gamma chain; Ck2b; Kiaa4163; proline rich calmodulin-dependent protein kinase; RATCAMKI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108058, 12323, 12325, 171140, 24245, 24246, 816, 817, 818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08413, P11730, P15791, P28652, Q13554, Q13555, Q13557, Q6PHZ2, Q923T9 | |
| Camk2b, CAMK2D, Camk2g | |
| Synthetic peptide corresponding to amino acids 248-299of human CaMKII beta. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction