missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKII gamma Monoclonal Antibody (8G10C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531665
This item is not returnable.
View return policy
Description
MA5-31665 has been tested in indirect ELISA.
Calcium/calmodulin-dependent protein kinase that functions autonomously after Ca(2+)/calmodulin-binding and autophosphorylation, and is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca(2+) transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner. This structural function is required for correct targeting of CaMK2A, which acts downstream of NMDAR to promote dendritic spine and synapse formation and maintain synaptic plasticity which enables long-term potentiation (LTP) and hippocampus-dependent learning. In developing hippocampal neurons, promotes arborization of the dendritic tree and in mature neurons, promotes dendritic remodeling. Participates in the modulation of skeletal muscle function in response to exercise. In slow-twitch muscles, is involved in regulation of sarcoplasmic reticulum (SR) Ca(2+) transport and in fast-twitch muscle participates in the control of Ca(2+) release from the SR through phosphorylation of triadin, a ryanodine receptor-coupling factor, and phospholamban (PLN/PLB), an endogenous inhibitor of SERCA2A/ATP2A2.
Specifications
| CaMKII gamma | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P11730, Q13555 | |
| CAMK2G | |
| Purified recombinant fragment of human CAMK2G (AA: 322-481) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 8G10C1 | |
| Unconjugated | |
| CAMK2G | |
| Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK2; CAMK2B; CAMK2D; Camk2g; CAMKB; CAMKD; CAMKG; CAMK-II; caMK-II subunit gamma; caM-kinase II gamma chain | |
| Mouse | |
| Protein G | |
| RUO | |
| 171140, 818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction