missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKK alpha Antibody (4A11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084254-M04
This item is not returnable.
View return policy
Description
CaMKK alpha Monoclonal antibody specifically detects CaMKK alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| CaMKK alpha | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| calcium/calmodulin-dependent protein kinase kinase 1, calcium/calmodulin-dependent protein kinase kinase 1, alpha, Calcium/calmodulin-dependent protein kinase kinase alpha, CaM-kinase IV kinase, CaM-kinase kinase 1, CaM-kinase kinase alpha, CaM-KK 1, CaMKK alpha, CaM-KK alpha, CAMKK alpha protein, CAMKKACaMKK 1, DKFZp761M0423, EC 2.7.11, EC 2.7.11.17, MGC34095 | |
| CAMKK1 (AAH31647.1, 2 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EGGPAVCCQDPRAELVERVAAIDVTHLEEADGGPEPTRNGVDPPPRARAASVIPGSTSRLLPARPSLSARKLSLQERPAGSYLEAQAGPYATGPASHIS | |
| 0.1 mg | |
| Protein Kinase | |
| 84254 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 4A11 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| AAH31647 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction