missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKK alpha Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42682AF700
This item is not returnable.
View return policy
Description
CaMKK alpha Polyclonal antibody specifically detects CaMKK alpha in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CaMKK alpha | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| calcium/calmodulin-dependent protein kinase kinase 1, calcium/calmodulin-dependent protein kinase kinase 1, alpha, Calcium/calmodulin-dependent protein kinase kinase alpha, CaM-kinase IV kinase, CaM-kinase kinase 1, CaM-kinase kinase alpha, CaM-KK 1, CaMKK alpha, CaM-KK alpha, CAMKK alpha protein, CAMKKACaMKK 1, DKFZp761M0423, EC 2.7.11, EC 2.7.11.17, MGC34095 | |
| The immunogen for this product maps to a region between residue 150 and 200 of human calcium/calmodulin-dependent protein kinase kinase 1, alpha using the numbering given in entry NP_757343.1 (GeneID 84254). | |
| 0.1 mL | |
| Protein Kinase | |
| 84254 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction