missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKV Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117829
This item is not returnable.
View return policy
Description
CaMKV Polyclonal Antibody for Western Blot, ICC/IF, IP
CaMKV does not appear to have detectable kinase activity.
Specifications
| CaMKV | |
| Polyclonal | |
| Unconjugated | |
| CAMKV | |
| 1G5; 1G5z; BB074618; BC017634; calcium/calmodulin dependent protein; CaM kinase like vesicle associated; CaM kinase-like vesicle-associated; CaM kinase-like vesicle-associated b; caM kinase-like vesicle-associated protein; CAMK5; Camkv; camkvb; DKFZp564G2082; hypothetical protein MGC63506; testis tissue sperm-binding protein Li 52e; VACAMKL; vesicle-associated calmodulin-binding protein; zgc:63506 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79012 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.24 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q8NCB2 | |
| CAMKV | |
| E. coli-derived Human CAMKV fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction