missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMLG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562655
This item is not returnable.
View return policy
Description
Immunogen sequence: GDKLDSFIKP PECSSDVNLE LRQRNRGDLT ADSVQRGSRH GLEQYLSRFE EAMKLRKQLI SEKPSQEDGN TTEE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.
The immunosuppressant drug cyclosporin A blocks a calcium-dependent signal from the T-cell receptor (TCR) that normally leads to T-cell activation. When bound to cyclophilin B, cyclosporin A binds and inactivates the key signaling intermediate calcineurin. The protein encoded by this gene functions similarly to cyclosporin A, binding to cyclophilin B and acting downstream of the TCR and upstream of calcineurin by causing an influx of calcium. This integral membrane protein appears to be a new participant in the calcium signal transduction pathway, implicating cyclophilin B in calcium signaling, even in the absence of cyclosporin.
Specifications
| CAMLG | |
| Polyclonal | |
| Unconjugated | |
| CAMLG | |
| AI385748; calcium modulating ligand; calcium signal-modulating cyclophilin ligand; calcium-modulating cyclophilin ligand; calcium-signal modulating cyclophilin ligand; CAML; Camlg; cyclophilin B-binding protein; GET2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 819 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49069 | |
| CAMLG | |
| Recombinant protein corresponding to Human CAMLG. Recombinant protein control fragment (Product #RP-101677). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction