Learn More
R&D Systems™ Camostat mesylate
Product Code. 18712614
Modifier l'affichage
Click to view available options
Quantity:
10 mg
50 mg
Conditionnement:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
18712614 10 mg 10mg
18782494 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18712614 Fournisseur R&D Systems™ Code fournisseur 3193/10
 Restricted Item
Estimated Shipment: 05-11-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
A Tocris product, part of R&D Systems. Orally active protease inhibitor
Camostat mesylate is an orally active protease inhibitor. Inhibits trypsin and various inflammatory proteases including plasmin, kallikrein and thrombin. Suppresses pancreatitis-induced pain in rats following oral administration. Also inhibits transmembrane serine protease TMPRSS2 and partially blocks entry of SARS-Cov-2 (COVID-19) into lung cells in vitro.
R&D Systems products are for biomedical research use only. They are not intended for human or veterinary use.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 59721-29-8
Molecular Formula C21H26N4O8S
Spécification
Quantity 10 mg
Product Line Tocris
Formula Weight 494.52
Purity >98%
Molecular Weight (g/mol) 494.52
Chemical Name or Material Camostat mesylate