missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ CHO-K1 CXCR2 (IL8RB) Gi Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The cAMP Hunter CXCR2 (IL8RB) Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR2 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype CXCR2 , a naturally Gi/Go coupled GPCR. Upon activation of CXCR2 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Intracellular cAMP level |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Recombinant Human IL-8 (CXCL8) |
| Quantity | 2 Vials |
| Target Entry | CXCR2 (IL8RB) |
| Accession Number | NM_001557 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction