missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ CHO-K1 GPBAR1 Bioassay Kit
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The cAMP Hunter GPBAR1 Bioassay kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This bioassay assesses ligand (e.g. peptide YY) based activation of GPBAR1 GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GPBAR1, a naturally Gi coupled GPCR. Upon activation of GPBAR1 , the (decrease/increase) in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Intracellular cAMP level |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Format | 1,000 dp (10 x 96-well) |
| Quantity | 1,000 dp (10 x 96-well) |
| Cell Line | CHO-K1 |
| Target | GPBAR1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction