missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Fisher Scientific™ Campylobacter jejuni ATCC 29428 5pcs
Click to view available options
Quantity:
5 Loops
Unit Size:
Pack of 5
Description
Recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | ATCC - EULA required |
| Biosafety Level | DG |
| Content And Storage | -18°C |
| Country of Origin | US |
| Product Type | Quality Control Organism |
| For Use With (Application) | Ready-to-Use ATCC QC Organisms |
| No. per Pack | 5 |
| Packaging Type | Foil Pk. |
| Quantity | 5 Loops |
| Species | Campylobacter jejuni |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction