missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMTA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578440
This item is not returnable.
View return policy
Description
Positive Control: human CAMTA1-transfected 293T Predicted Reactivity: Mouse (88%), Rat (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMTA1 is a transcriptional activator. May act as a tumor suppressor.
Specifications
| CAMTA1 | |
| Polyclonal | |
| Unconjugated | |
| CAMTA1 | |
| 1810059M14Rik; 2310058O09Rik; AI316882; AW987475; calmodulin binding transcription activator 1; calmodulin-binding transcription activator 1; Camta1; CANPMR; Kiaa0833; mKIAA0833; MSTP023 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23261 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.0 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9Y6Y1 | |
| CAMTA1 | |
| Recombinant protein encompassing a sequence within the center region of human CAMTA1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction