missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAND1 Polyclonal Antibody

Product Code. 15761832 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15761832 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15761832 Supplier Invitrogen™ Supplier No. PA564066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QTRPVQSWLC DPDAMEQGET PLTMLQSQVP NIVKALHKQM KEKSVKTRQC CFNMLTELVN VLPG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

CAND1 enhances transcription from various types of promoters. It is a regulatory protein that interferes with the assembly of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex and thereby down-regulates ubiquitination of target proteins. CAND1 prevents neddylation of CUL1 by physically blocking access to the neddylation site. CAND1 disrupts interactions between CUL1 and SKP1A and between CUL1 and F-box proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAND1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAND1
Gene Accession No. Q86VP6
Gene Alias 2310038O07Rik; 6330512O03Rik; AI195005; AI846556; CAND1; cullin associated and neddylation disassociated 1; cullin associated and neddylation dissociated 1; cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated protein 1; Cullin-associated NEDD8-dissociated protein 1; D10Ertd516e; DKFZp434M1414; fb78a05; FLJ10114; FLJ10929; FLJ38691; FLJ90441; KIAA0829; mKIAA0829; p120 CAND1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120A; TBP-interacting protein of 120 kDa A; TBP-interacting protein TIP120A; TIP120; TIP120 protein; TIP120A; Tp120a; wu:fb78a05; zgc:55729
Gene Symbols CAND1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAND1. Recombinant protein control fragment (Product #RP-103473).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.