missing translation for 'onlineSavingsMsg'
Learn More

Canertinib, MedChemExpress

Product Code. p-200100838
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16490976

Brand: Medchem Express HY10367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Canertinib (CI-1033;PD-183805) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Canertinib is active against vaccinia virus respiratory infection in mice.

Canertinib (CI-1033

Specifications

Chemical Name or Material Canertinib
CAS 267243-28-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H25ClFN5O3
Quantity 200 mg
Synonym CI-1033 PD-183805
Solubility Information Ethanol : 12.5 mg/mL (25.72 mM; Need ultrasonic) ∣DMSO : 4.9 mg/mL (10.08 mM; Need warming)
SMILES O=C(NC1=C(C=C2C(C(NC3=CC=C(C(Cl)=C3)F)=NC=N2)=C1)OCCCN4CCOCC4)C=C
Molecular Weight (g/mol) 485.94
Formula Weight 485.94
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.