missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine Distemper Virus Antibody (DV2-12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64816DL594
This item is not returnable.
View return policy
Description
Canine Distemper Virus Monoclonal antibody specifically detects Canine Distemper Virus in Canine, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Canine Distemper Virus | |
| Monoclonal | |
| DyLight 594 | |
| CDV | |
| Made to Viral CANINE DISTEMPER VIRUS | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DV2-12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Canine, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction