missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine HGFR/c-MET Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF4140
This item is not returnable.
View return policy
Description
HGFR/c-MET Polyclonal specifically detects HGFR/c-MET in Canine samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| HGFR/c-MET | |
| Polyclonal | |
| Biotin | |
| Q75ZY9 | |
| MET | |
| Mouse myeloma cell line NS0-derived recombinant canine HGF R Glu25-Leu935 Accession # Q75ZY9 | |
| 50 μg | |
| Primary | |
| Detects canine HGF R in ELISAs and Western blots. In sandwich immunoassays, less than 7% cross-reactivity with recombinant mouse HGF R is observed and less than 0.2% cross-reactivity with recombinant human HGF R is observed. | |
| Canine | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4233 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction