missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems BAF754
This item is not returnable.
View return policy
Description
IL-4 Polyclonal specifically detects IL-4 in Canine samples. It is validated for Western Blot.
Specifications
| IL-4 | |
| Polyclonal | |
| Biotin | |
| AAD11563 | |
| IL4 | |
| E. coli-derived recombinant canine IL-4 (R&D Systems, Catalog # 754-CL) His25-His132 Accession # AAD11563 | |
| 50 μg | |
| Primary | |
| Detects canine IL-4 in Western blots. In this format, approximately 25% cross-reactivity with rhIL-4 is observed, 10% cross-reactivity with rpIL-4 is observed and less than 1% cross-reactivity with rmIL-4 and rrIL-4 is observed. | |
| Canine | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3565 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction