missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1609-SP
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Canine samples. It is validated for Neutralization.
Specifications
| IL-6 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 10-30 ug/mL | |
| P41323 | |
| IL6 | |
| E. coli-derived recombinant canine IL-6 Thr23-Met207 Accession # P41323 | |
| 25 μg | |
| Primary | |
| Detects canine IL-6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Cardiotrophin-1, recombinant human (rh) CLC, recombinant rat (rr) CNTF, recombinant cotton rat IL-6, recombinant equine IL-6, rhIL-6, recombinant porcine IL-6, rrIL-6, rmIL-11, rmLIF, or rmOncostatin M is observed. | |
| Canine | |
| Purified |
| Neutralization | |
| 247017 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3569 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction