missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Canine IL-6 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
Description
The Canine IL-6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies canine IL-6.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Canine |
| Conjugate | Biotin |
| Assay Range | 62.50 to 4000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 403985 |
| Synonym | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction