missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-8/CXCL8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB16081-SP
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Canine samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 3-12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P41324 | |
| IL8 | |
| E. coli-derived recombinant canine IL-8/CXCL8 Ala23-Pro101 Accession # P41324 | |
| 25 μg | |
| Primary | |
| Detects canine IL-8/CXCL8 in ELISAs. In ELISAs, no cross-reactivity with recombinant feline IL-8/CXCL8, recombinant human CXCL8/IL-8, recombinant porcine CXCL8/IL-8, recombinant canine (rca) IL-1 beta/IL-1F2, or rcaCXCL6/IL-6 is observed. | |
| Canine | |
| Purified |
| Neutralization, ELISA | |
| 258901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3576 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction