missing translation for 'onlineSavingsMsg'
Learn More

Canine Parvovirus Antibody (2G10A3) - BSA Free, Novus Biologicals™

Product Code. p-200080623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042586 0.1 mL 0.1mL
18693917 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18042586 Supplier Novus Biologicals Supplier No. NBP261721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Canine Parvovirus Monoclonal specifically detects Canine Parvovirus in Human, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Parvovirus
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2G10A3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 -1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias CPV, Parvo
Host Species Mouse
Immunogen Purified recombinant fragment of Canine Parvovirus CPV-VP2 (AA: 1-128) expressed in E. Coli.
Molecular Weight of Antigen 68.6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.