missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine Parvovirus Antibody (7341), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62433
This item is not returnable.
View return policy
Description
Canine Parvovirus Monoclonal specifically detects Canine Parvovirus in Virus samples. It is validated for ELISA.
Specifications
| Canine Parvovirus | |
| Monoclonal | |
| Unconjugated | |
| CPV, Parvo | |
| Canine parvovirus | |
| 0.05 mg | |
| Primary | |
| Virus | |
| IgG2a |
| ELISA | |
| 7341 | |
| ELISA 1:20-1:200 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| This product detects Canine parvovirus (CPV), a negatively stranded DNA virus which infects dogs. In some instances it is also known as CPV type-2 to distinguish it from the unrelated parvovirus minute virus of canines (MVC). CPV is a relatively new virus, having been first isolated in 1978, which is thought to have derived from the feline panleukopenia virus (FPV). CPV is highly infectious and is spread from dog to dog by physical contact and contact with feces. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction