Learn More
Invitrogen™ Canine Parvovirus Monoclonal Antibody (7354)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547586
Description
Specificity: Reactive with CPV. Non-reactive with Feline Panleukopenia virus. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Canine parvovirus is a contagious virus mainly affecting dogs. CPV is highly contagious and is spread from dog to dog by direct or indirect contact with their feces. Vaccines can prevent this infection, but mortality can reach 91% in untreated cases. Treatment often involves veterinary hospitalization.
Specifications
| Canine Parvovirus | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 7354 | |
| Unconjugated | |
| CPV; parvo | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C or -20°C if preferred | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.