missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine VEGF164 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1603
This item is not returnable.
View return policy
Description
VEGF Polyclonal specifically detects VEGF in Canine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| VEGF | |
| Polyclonal | |
| Unconjugated | |
| Q9MYV3 | |
| VEGFA | |
| E. coli-derived recombinant canine VEGF 164 Pro28-Arg190 Accession # Q9MYV3 | |
| 100 μg | |
| Primary | |
| Detects canine VEGF 164 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human VEGF, recombinant mouse VEGF, and recombinant rat VEGF is observed. | |
| Canine | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.04-0.10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7422 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction