Learn More
Bode Chemie™ Canister-Key
Product Code. 10323221
Change view
Click to view available options
For Use With (Equipment):
BODE 2L, 5L and 10L Canister
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10323221 BODE 2L, 5L and 10L Canister Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10323221 Supplier Bode Chemie™ Supplier No. 9800510
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Canister-Key
For Use With (Equipment) BODE 2L, 5L and 10L Canister
No. per Pack 1 Ea.