missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody (2A1), Novus Biologicals™
Human Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Cannabinoid R1/CB1/CNR1 |
|---|---|
| Clone | 2A1 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043032
|
Novus Biologicals
NBP3-26648-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043836
|
Novus Biologicals
NBP3-26648-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for ELISASpecifications
| Cannabinoid R1/CB1/CNR1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
| Recombinant Human Cannabinoid R1/CB1/CNR1 protein [UniProt P21554] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2A1 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 1268 | |
| IgG4 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title