missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R2/CB2/CNR2 Antibody (352110R), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36551RMFV450
This item is not returnable.
View return policy
Description
Cannabinoid R2/CB2/CNR2 Monoclonal antibody specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Cannabinoid R2/CB2/CNR2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Neuroscience, Vision | |
| 1269 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 352110R | |
| Flow Cytometry, CyTOF-ready | |
| cannabinoid receptor 2 (macrophage), CB-2, CB2cannabinoid receptor 2, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
| Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction