missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cannabinoid Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16392025
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16392025 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16392025 Supplier Invitrogen™ Supplier No. PA585080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Cat (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Positive Control: U87-MG, human CB1-transfected HEK-293T cells, mouse brain, Rat2, NSC34. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cannabinoid Receptor 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CNR1
Gene Accession No. P21554, P47746
Gene Alias brain-type cannabinoid receptor; CANN6; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB1; CB1 cannabinoid receptor; CB1 receptor; CB1A; CB1K5; CB1R; CB-R; central cannabinoid receptor; CNR; Cnr1; HGNC:2159; OTTHUMP00000016839; RP1-23D17.1; seven transmembrane domains; G-protein-coupled receptor; Skr6; SKR6R; striatal cannabinoid receptor type 1 protein
Gene Symbols CNR1
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human Cannabinoid Receptor 1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1268, 12801
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.