missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cannabinoid Receptor 1 Recombinant Human Monoclonal Antibody (2A1)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550075
This item is not returnable.
View return policy
Description
Cannabinoid Receptor 1 Recombinant Monoclonal Antibody for ELISA
This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.
Specifications
| Cannabinoid Receptor 1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P21554 | |
| CNR1 | |
| Recombinant Human CNR1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG4 |
| ELISA, Neutralization | |
| 2A1 | |
| Unconjugated | |
| CNR1 | |
| brain-type cannabinoid receptor; CANN6; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB1; CB1 cannabinoid receptor; CB1 receptor; CB1A; CB1K5; CB1R; CB-R; central cannabinoid receptor; CNR; Cnr1; HGNC:2159; OTTHUMP00000016839; RP1-23D17.1; seven transmembrane domains; G-protein-coupled receptor; Skr6; SKR6R; striatal cannabinoid receptor type 1 protein | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 1268 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction