Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ CFR08NC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15999999
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
100 to 500 cSt
0.5
ASTM D 445, D 446 and ISO 3104, 3105
No
D 445, D 446
350
Certificates
Special Offers
Special Offers
4 week lead-time