missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CANT1 Mouse anti-Human, Clone: OTI7F8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805629
This item is not returnable.
View return policy
Description
This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.Specifications
| CANT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WVQ1 | |
| CANT1 | |
| Human recombit protein fragment corresponding to amino acids 63-401 of human CANT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7F8 | |
| Unconjugated | |
| CANT1 | |
| DBQD, DBQD1, EDM7, SCAN-1, SCAN1, SHAPY | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 124583 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction