missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CANT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554612
This item is not returnable.
View return policy
Description
Immunogen sequence: TTTTGDVVNE NPEWVKVVGY KGSVDHENWV SNYNALRAAA GIQPPGYLIH ESACWSDTLQ RWFFLPRRAS QERYSEKDD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.
Specifications
| CANT1 | |
| Polyclonal | |
| Unconjugated | |
| CANT1 | |
| 5830420C20Rik; Apy1h; apyrase 1, homolog (C. lectularius); apyrase homolog; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; calcium activated nucleotidase 1; Cant1; D11Bwg0554e; DBQD; DBQD1; EC 3.6.1.6; ectonucleoside triphosphate diphosphohydrolase 8; Entpd8; galectin-3-binding protein; soluble calcium-activated nucleotidase 1; lectin, galactoside-binding, soluble, 3 binding protein; LGALS3BP; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN1; SCAN-1; SHAPY; soluble Ca-activated nucleotidase, isozyme 1; soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1; srapy | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124583 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WVQ1 | |
| CANT1 | |
| Recombinant protein corresponding to Human CANT1. Recombinant protein control fragment (Product #RP-92530). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction