missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CANT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16538301
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16538301 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16538301 Supplier Invitrogen™ Supplier No. PA592914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HRPAPGRPPT HNAHNWRLGQ APANWYNDTY PLSPPQRTPA GIRYRIAVIA DLDTESRAQE ENTWFSYLKK GYLTLSDSGD KVAVEWDKDH GVLESHLAEK GRGMELSDLI VFNGKLYSVD DRTGVVYQIE GSKAVPWVIL SDGDGTVEKG FKAEWLAVKD ERLYVGGLGK Positive Samples: U-251MG; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein.

This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CANT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CANT1
Gene Accession No. Q8WVQ1
Gene Alias 5830420C20Rik; Apy1h; apyrase 1, homolog (C. lectularius); apyrase homolog; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; calcium activated nucleotidase 1; Cant1; D11Bwg0554e; DBQD; DBQD1; EC 3.6.1.6; ectonucleoside triphosphate diphosphohydrolase 8; Entpd8; galectin-3-binding protein; soluble calcium-activated nucleotidase 1; lectin, galactoside-binding, soluble, 3 binding protein; LGALS3BP; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN1; SCAN-1; SHAPY; soluble Ca-activated nucleotidase, isozyme 1; soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1; srapy
Gene Symbols CANT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 62-231 of human CANT1 (NP_6201481).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124583
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.