missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAP1 Antibody (SR1982), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22480-50ul
This item is not returnable.
View return policy
Description
CAP1 Monoclonal antibody specifically detects CAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| CAP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1982 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| adenylyl cyclase-associated protein 1, CAP, adenylate cyclase-associated protein 1 (yeast), CAP1-PEN, CAPCAP 1, SSKAP55 | |
| A synthesized peptide derived from human CAP1 (Uniprot #: Q01518) | |
| 50 μg | |
| Signal Transduction | |
| 10487 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction