missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAP1 Recombinant Rabbit Monoclonal Antibody (JE61-08)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544851
This item is not returnable.
View return policy
Description
CAP1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.
Specifications
| CAP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q01518 | |
| CAP1 | |
| Synthetic peptide within human CAP1 aa 10-59/475. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE61-08 | |
| Unconjugated | |
| CAP1 | |
| adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction