missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CapG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 226.00 - € 481.00
Specifications
| Antigen | CapG |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687180
|
Novus Biologicals
NBP2-92955-0.02ml |
0.02 mL |
€ 226.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656951
|
Novus Biologicals
NBP2-92955-0.1ml |
0.1 mL |
€ 481.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CapG Polyclonal antibody specifically detects CapG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-OutSpecifications
| CapG | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 822 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-348 of human CAPG (NP_001738.2). MYTAIPQSGSPFPGSVQDPGLHVWRVEKLKPVPVAQENQGVFFSGDSYLVLHNGPEEVSHLHLWIGQQSSRDEQGACAVLAVHLNTLLGERPVQHREVQGNESDLFMSYFPRGLKYQEGGVESAFHKTSTGAPAAIKKLYQVKGKKNIRATERALNWDSFNTGDCFILDLGQNIFAWCGGKSNILERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGHESPIFKQFFKDWK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title